The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027259.1 | NP_081535.1 | 1 - 2 | 330 | 77 |
GenBank mRNA | AB221537.1 | - | 1 - 2 | 58 | 77 |
BC062812.1 | - | 1 - 2 | 330 | 77 |