Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smg9
Entrez Gene ID
71997
Gene Name
smg-9 homolog, nonsense mediated mRNA decay factor (C. elegans)
Gene Aliases
1500002O20Rik, AI429693, N28092
Chromosome Location
Chr.7: 24398910 - 24422777 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028047.2 NP_082323.1 13 - 14 1758 69
GenBank mRNA AK005126.1 - 13 - 14 1758 69
AK141576.1 - 13 - 14 1749 69
AK169651.1 - 13 - 14 1682 69
BC034211.1 - 13 - 14 1673 69

View Full Product Details