Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fez1
Entrez Gene ID
235180
Gene Name
fasciculation and elongation protein zeta 1 (zygin I)
Gene Aliases
UNC-76, UNC76
Chromosome Location
Chr.9: 36821944 - 36878926 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK138656.1 - 1 - 2 232 74
BC029629.1 - 1 - 2 112 74

View Full Product Details