The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145960.1 | NP_001139432.1 | 10 - 11 | 1359 | 60 |
NM_020258.4 | NP_064654.3 | 10 - 11 | 1359 | 60 | |
GenBank mRNA | AF121081.1 | - | 10 - 11 | 1258 | 60 |
AK028984.1 | - | 10 - 11 | 1035 | 60 | |
AK031045.1 | - | 10 - 11 | 1297 | 60 | |
AK079414.1 | - | 10 - 11 | 1035 | 60 | |
AK079823.1 | - | 10 - 11 | 1360 | 60 | |
AK220151.1 | - | 8 - 9 | 801 | 60 | |
BC038000.1 | - | 10 - 11 | 1030 | 60 | |
BC063326.1 | - | 10 - 11 | 1047 | 60 |