The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145960.1 | NP_001139432.1 | 6 - 7 | 912 | 60 |
NM_020258.4 | NP_064654.3 | 6 - 7 | 912 | 60 | |
GenBank mRNA | AF121081.1 | - | 6 - 7 | 811 | 60 |
AK028984.1 | - | 6 - 7 | 588 | 60 | |
AK031045.1 | - | 6 - 7 | 850 | 60 | |
AK079414.1 | - | 6 - 7 | 588 | 60 | |
AK079823.1 | - | 6 - 7 | 913 | 60 | |
AK220151.1 | - | 4 - 5 | 354 | 60 | |
BC038000.1 | - | 6 - 7 | 583 | 60 | |
BC063326.1 | - | 6 - 7 | 600 | 60 |