Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Zbtb16
- Entrez Gene ID
-
235320
- Gene Name
- zinc finger and BTB domain containing 16
- Gene Aliases
-
AI467657, PLZF, Zfp145, lu
- Chromosome Location
-
Chr.9: 48651797 - 48836381 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
Z47205.1 |
- |
1 - 2 |
200 |
93 |
View Full Product Details