Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lman1l
Entrez Gene ID
235416
Gene Name
lectin, mannose-binding 1 like
Gene Aliases
BC020188, slamp
Chromosome Location
Chr.9: 57606717 - 57621419 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199222.3 NP_954692.2 3 - 4 547 67
GenBank mRNA AB211986.1 - 3 - 4 468 67
BC020188.1 - 3 - 4 558 67
BY708954.1 - 3 - 4 548 67

View Full Product Details