The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034085.2 | NP_001029257.1 | 1 - 2 | 170 | 80 |
NM_001286553.1 | NP_001273482.1 | 1 - 2 | 170 | 80 | |
NM_028614.3 | NP_082890.2 | 1 - 2 | 170 | 80 | |
GenBank mRNA | AK077158.1 | - | 1 - 2 | 124 | 80 |
AK136106.1 | - | 1 - 2 | 128 | 80 | |
AK161392.1 | - | 1 - 2 | 123 | 80 | |
BC056218.1 | - | 1 - 2 | 164 | 80 | |
BY722866.1 | - | 1 - 2 | 171 | 80 |