The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034085.2 | NP_001029257.1 | 4 - 5 | 601 | 69 |
NM_001286553.1 | NP_001273482.1 | 4 - 5 | 601 | 69 | |
NM_028614.3 | NP_082890.2 | 4 - 5 | 601 | 69 | |
GenBank mRNA | AK077158.1 | - | 4 - 5 | 555 | 69 |
AK136106.1 | - | 4 - 5 | 559 | 69 | |
AK161392.1 | - | 4 - 5 | 554 | 69 | |
BC056218.1 | - | 4 - 5 | 595 | 69 |