Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jrkl
Entrez Gene ID
77532
Gene Name
Jrk-like
Gene Aliases
C030035D04Rik, HHMJG
Chromosome Location
Chr.9: 13242790 - 13245741 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033181.1 NP_001028353.1 1 - 1 2825 134
GenBank mRNA AK079877.1 - 1 - 1 1950 134
BB210598.2 - 1 - 1 530 134

View Full Product Details