Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx1
Entrez Gene ID
56440
Gene Name
sorting nexin 1
Gene Aliases
-
Chromosome Location
Chr.9: 66088127 - 66124886 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019727.2 NP_062701.2 10 - 11 1134 63
GenBank mRNA AB019214.1 - 10 - 11 1074 63
AF154120.1 - 10 - 11 1138 63
AK076198.1 - 2 - 3 119 63
AK149850.1 - 10 - 11 1046 63
AK154079.1 - 10 - 11 1047 63
AK155746.1 - 10 - 11 1022 63
AK161241.1 - 10 - 11 1115 63
AK167177.1 - 10 - 11 1089 63
AK167993.1 - 10 - 11 1029 63
AK168742.1 - 10 - 11 1127 63
BC066044.1 - 10 - 11 1091 63
BC066189.1 - 10 - 11 1025 63

View Full Product Details