The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009022.4 | NP_033048.2 | 3 - 4 | 422 | 82 |
GenBank mRNA | AK078553.1 | - | 3 - 4 | 363 | 82 |
BC075704.1 | - | 3 - 4 | 389 | 82 | |
CJ166631.1 | - | 3 - 4 | 423 | 82 |