The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198962.3 | NP_945200.1 | 7 - 8 | 1394 | 85 |
GenBank mRNA | AK038551.1 | - | 7 - 8 | 1395 | 85 |
AK048781.1 | - | 7 - 8 | 1394 | 85 | |
AK079572.1 | - | 7 - 8 | 1428 | 85 | |
AY336085.1 | - | 7 - 8 | 1362 | 85 | |
BC140957.1 | - | 7 - 8 | 1390 | 85 |