Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stoml1
Entrez Gene ID
69106
Gene Name
stomatin-like 1
Gene Aliases
1810015E19Rik, AI847431, SLP1, STORP, UNC24, WPB72
Chromosome Location
Chr.9: 58253164 - 58262524 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026942.3 NP_081218.3 3 - 4 495 55
NR_028146.1 4 - 5 775 55
GenBank mRNA AK007508.1 - 1 - 2 164 55
AK148324.1 - - 394 55
BC017536.1 - 4 - 5 747 55
BC037074.1 - 3 - 4 425 55

View Full Product Details