Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Opcml
- Entrez Gene ID
-
330908
- Gene Name
- opioid binding protein/cell adhesion molecule-like
- Gene Aliases
-
2900075O15Rik, 3732419F12, AI844366, B930023M13Rik, C230027C17, Gm181, Obcam
- Chromosome Location
-
Chr.9: 27790952 - 28925410 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_177906.4 |
NP_808574.2 |
7 - 8 |
1036 |
64 |
GenBank mRNA |
BC076581.1 |
- |
7 - 8 |
1036 |
64 |
View Full Product Details