Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Serinc1
Entrez Gene ID
56442
Gene Name
serine incorporator 1
Gene Aliases
1500011D18Rik, AI315070, AIGP-2, TMS-2, Tde1l, Tde2, Tms2, mKIAA1253
Chromosome Location
Chr.10: 57515774 - 57532529 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019760.3 NP_062734.1 7 - 8 1129 117
GenBank mRNA AB029500.1 - 7 - 8 1073 117
AF181685.1 - 7 - 8 1049 117
AK002847.1 - 7 - 8 1078 117
AK005203.1 - 7 - 8 1073 117
AK078632.1 - 1 - 2 78 117
AK088340.1 - 7 - 8 1071 117
AK133668.1 - 7 - 8 1070 117
AK133975.1 - 7 - 8 1070 117
AK135359.1 - 7 - 8 1047 117
AK141574.1 - 7 - 8 1078 117
AK148745.1 - 7 - 8 1050 117
AK159280.1 - 7 - 8 1044 117
AK159436.1 - 7 - 8 1096 117
AK167817.1 - 7 - 8 1077 117
BC006940.1 - 7 - 8 1102 117
BC017148.1 - 7 - 8 1019 117

View Full Product Details