The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_205823.2 | NP_991392.1 | 1 - 1 | 3116 | 76 |
GenBank mRNA | AK137624.1 | - | 1 - 1 | 3117 | 76 |