The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011260.1 | NP_035390.1 | 3 - 4 | 232 | 102 |
GenBank mRNA | AK008446.1 | - | 3 - 4 | 228 | 102 |
AK008608.1 | - | 3 - 4 | 228 | 102 | |
BC061139.1 | - | 3 - 4 | 230 | 102 | |
D63361.1 | - | 3 - 4 | 232 | 102 |