Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Reg3a
Entrez Gene ID
19694
Gene Name
regenerating islet-derived 3 alpha
Gene Aliases
AV063448
Chromosome Location
Chr.6: 78380709 - 78383839 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011259.1 NP_035389.1 1 - 2 73 82
GenBank mRNA BC119153.1 - 1 - 2 73 82
D63356.1 - 1 - 2 73 82

View Full Product Details