The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011259.1 | NP_035389.1 | 5 - 6 | 537 | 63 |
GenBank mRNA | AK008050.1 | - | 2 - 3 | 192 | 63 |
AK008140.1 | - | 2 - 3 | 192 | 63 | |
AK008470.1 | - | 3 - 4 | 372 | 63 | |
BC119153.1 | - | 5 - 6 | 537 | 63 | |
D63356.1 | - | 5 - 6 | 537 | 63 | |
D63357.1 | - | 5 - 6 | 486 | 63 |