Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rmnd5a
Entrez Gene ID
68477
Gene Name
required for meiotic nuclear division 5 homolog A (S. cerevisiae)
Gene Aliases
1110007A06Rik, AJ237586, AW125795, Gid2, MTA.D02.090
Chromosome Location
Chr.6: 71388634 - 71440637 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024288.2 NP_077250.2 7 - 8 1452 91
GenBank mRNA AK148556.1 - 6 - 7 687 91
BC034085.1 - 3 - 4 438 91
BC050876.1 - 7 - 8 1303 91

View Full Product Details