Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Crbn
Entrez Gene ID
58799
Gene Name
cereblon
Gene Aliases
2610203G15Rik, 2900045O07Rik, AF229032, AW108261, piL
Chromosome Location
Chr.6: 106778244 - 106800087 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021449.3 NP_067424.2 8 - 9 984 121
NM_175357.3 NP_780566.1 8 - 9 987 121
GenBank mRNA AF229032.1 - 8 - 9 939 121
AK032836.1 - 8 - 9 961 121
AK050557.1 - 8 - 9 970 121
AK075959.1 - 8 - 9 960 121
AK076144.1 - 8 - 9 967 121
AK160219.1 - 8 - 9 963 121
BC046967.1 - 7 - 8 902 121
BC069905.1 - 8 - 9 944 121
BC086488.1 - 8 - 9 962 121

View Full Product Details