Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rab11fip5
Entrez Gene ID
52055
Gene Name
RAB11 family interacting protein 5 (class I)
Gene Aliases
9130206P09Rik, C75969, D6Ertd32e, GAF1, RIP11
Chromosome Location
Chr.6: 85334962 - 85374648 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003955.2 NP_001003955.1 5 - 6 3866 96
NM_177466.4 NP_803417.3 4 - 5 1847 96
GenBank mRNA AK129230.1 - 5 - 6 3866 96
AK145061.1 - 4 - 5 1360 96
AK154922.1 - 5 - 6 3851 96
BC026473.1 - 2 - 3 537 96
BC044833.1 - 4 - 5 1376 96
BC051063.1 - 4 - 5 1376 96
BC141380.1 - 4 - 5 1777 96

View Full Product Details