Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tprkb
Entrez Gene ID
69786
Gene Name
Tp53rk binding protein
Gene Aliases
0610033G21Rik, 1810034M08Rik, Cgi-121, DRWMS1, mDRWMS1
Chromosome Location
Chr.6: 85915278 - 85930284 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176842.3 NP_789812.1 1 - 2 75 89
GenBank mRNA BY108059.1 - 1 - 2 76 89

View Full Product Details