Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plekhg6
Entrez Gene ID
213522
Gene Name
pleckstrin homology domain containing, family G (with RhoGef domain) member 6
Gene Aliases
BC026778, Gm155
Chromosome Location
Chr.6: 125362651 - 125380951 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198604.2 NP_941006.2 15 - 16 2122 118
GenBank mRNA AK153619.1 - 15 - 16 2412 118
BC026778.1 - 15 - 16 2125 118

View Full Product Details