Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dyrk4
Entrez Gene ID
101320
Gene Name
dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 4
Gene Aliases
AW049118, Dyrk4a, Dyrk4b
Chromosome Location
Chr.6: 126876020 - 126921864 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207210.2 NP_997093.2 8 - 9 960 79
GenBank mRNA AK077117.1 - 8 - 9 961 79
BC052324.1 - 7 - 8 1014 79

View Full Product Details