Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cecr2
Entrez Gene ID
330409
Gene Name
cat eye syndrome chromosome region, candidate 2
Gene Aliases
2610101O16Rik, 2810409N01Rik, Gtl4, mKIAA1740
Chromosome Location
Chr.6: 120666320 - 120771194 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128151.1 NP_001121623.1 8 - 9 1114 79

View Full Product Details