Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klrk1
Entrez Gene ID
27007
Gene Name
killer cell lectin-like receptor subfamily K, member 1
Gene Aliases
D6H12S2489E, NKG2-D, Nkg2d
Chromosome Location
Chr.6: 129610323 - 129623864 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083322.2 NP_001076791.1 3 - 4 206 110
NM_001286018.1 NP_001272947.1 3 - 4 206 110
NM_033078.4 NP_149069.1 3 - 4 299 110
GenBank mRNA AA174479.1 - 3 - 4 315 110
AF030313.1 - 3 - 4 183 110
AF039026.1 - 3 - 4 213 110
AF054819.1 - 4 - 5 473 110
AK040436.1 - 3 - 4 288 110
AK170499.1 - 3 - 4 207 110
AY860972.1 - 3 - 4 189 110
BC057147.1 - 3 - 4 250 110
CT010384.1 - 3 - 4 189 110

View Full Product Details