Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec4n
Entrez Gene ID
56620
Gene Name
C-type lectin domain family 4, member n
Gene Aliases
Clec6a, Clecsf10, Nkcl
Chromosome Location
Chr.6: 123229843 - 123247024 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190320.1 NP_001177249.1 2 - 3 294 62
NM_020001.2 NP_064385.1 3 - 4 384 62
GenBank mRNA AF240357.1 - 3 - 4 353 62
AF240359.1 - 3 - 4 353 62
AK089166.1 - 3 - 4 371 62
AK131697.1 - 3 - 4 371 62
AK154673.1 - 2 - 3 281 62
AK171046.1 - 4 - 5 554 62
BC023008.1 - 3 - 4 353 62

View Full Product Details