The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027391.3 | NP_081667.1 | 4 - 5 | 695 | 109 |
GenBank mRNA | AK002363.1 | - | 4 - 5 | 696 | 109 |
BC023358.1 | - | 4 - 5 | 718 | 109 |