Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tsen2
Entrez Gene ID
381802
Gene Name
tRNA splicing endonuclease 2 homolog (S. cerevisiae)
Gene Aliases
AU067695
Chromosome Location
Chr.6: 115544704 - 115578340 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199033.1 NP_950198.1 2 - 3 229 75
GenBank mRNA AK134697.1 - 2 - 3 225 75
BC061473.1 - 2 - 3 229 75

View Full Product Details