Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BC048546
Entrez Gene ID
232400
Gene Name
cDNA sequence BC048546
Gene Aliases
Ovos
Chromosome Location
Chr.6: 128539822 - 128581606 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001179.3 NP_001001179.2 15 - 16 1903 95
GenBank mRNA AK138836.1 - 15 - 16 1904 95
AK161068.1 - 2 - 3 253 95
AK161184.1 - 15 - 16 1906 95
BC040781.1 - 3 - 4 355 95

View Full Product Details