The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001179.3 | NP_001001179.2 | 29 - 30 | 3818 | 91 |
GenBank mRNA | AK138836.1 | - | 29 - 30 | 3819 | 91 |
AK161068.1 | - | 16 - 17 | 2168 | 91 | |
AK161184.1 | - | 29 - 30 | 3822 | 91 | |
BC034132.1 | - | 4 - 5 | 578 | 91 | |
BC040781.1 | - | 17 - 18 | 2270 | 91 |