Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bms1
Entrez Gene ID
213895
Gene Name
BMS1 homolog, ribosome assembly protein (yeast)
Gene Aliases
AA408648, AU020092, AU043373, BB007109, BC030906, Bms1l, mKIAA0187
Chromosome Location
Chr.6: 118383381 - 118420018 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194339.1 NP_919320.1 15 - 16 2758 126
GenBank mRNA AK129082.1 - 15 - 16 2718 126
AK133013.1 - 15 - 16 2708 126
AK162178.1 - 15 - 16 2816 126
BC030906.1 - 8 - 9 1558 126
BC057054.1 - 15 - 16 2758 126

View Full Product Details