Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nagk
Entrez Gene ID
56174
Gene Name
N-acetylglucosamine kinase
Gene Aliases
Gnk
Chromosome Location
Chr.6: 83795031 - 83802556 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK007240.1 - 1 - 2 219 86

View Full Product Details