Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdca3
Entrez Gene ID
14793
Gene Name
cell division cycle associated 3
Gene Aliases
2410005A12Rik, C8, Grcc8, Tome-1
Chromosome Location
Chr.6: 124829595 - 124833701 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013538.5 NP_038566.1 1 - 2 95 96
GenBank mRNA AK008606.1 - 1 - 2 48 96
AK011313.1 - 1 - 2 95
BB860939.1 - 1 - 2 101 96

View Full Product Details