The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277330.1 | NP_001264259.1 | 10 - 11 | 1120 | 93 |
NM_001277805.1 | NP_001264734.1 | 11 - 12 | 1229 | 93 | |
NM_008995.2 | NP_033021.2 | 11 - 12 | 1192 | 93 | |
NM_175933.2 | NP_787947.1 | 10 - 11 | 1118 | 93 | |
GenBank mRNA | AJ416473.1 | - | 10 - 11 | 1100 | 93 |
AK088886.1 | - | 10 - 11 | 1122 | 93 | |
AK145111.1 | - | 11 - 12 | 1165 | 93 | |
AK145361.1 | - | 10 - 11 | 1055 | 93 | |
AK161470.1 | - | 10 - 11 | 1115 | 93 | |
BC029748.1 | - | 11 - 12 | 1199 | 93 |