Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ldlrap1
Entrez Gene ID
100017
Gene Name
low density lipoprotein receptor adaptor protein 1
Gene Aliases
AA691260, ARH2, Arh, Arh1, FHCB1, FHCB2
Chromosome Location
Chr.4: 134741554 - 134768004 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145554.2 NP_663529.2 8 - 9 884 61
GenBank mRNA AK148340.1 - 9 - 10 846 61
AK159800.1 - 8 - 9 828 61
AK167290.1 - 8 - 9 833 61
AK169675.1 - 3 - 4 317 61
BC021467.1 - 8 - 9 833 61
BC066808.1 - 8 - 9 827 61
BC067411.1 - 8 - 9 827 61

View Full Product Details