The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172416.3 | NP_766004.1 | 3 - 4 | 695 | 89 |
GenBank mRNA | AF533890.1 | - | 3 - 4 | 694 | 89 |
AK004546.1 | - | 3 - 4 | 672 | 89 | |
AK044897.1 | - | 3 - 4 | 694 | 89 | |
AK050513.1 | - | 3 - 4 | 669 | 89 | |
AK053664.1 | - | 3 - 4 | 645 | 89 | |
BC057635.1 | - | 3 - 4 | 633 | 89 |