Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrtm2
Entrez Gene ID
211187
Gene Name
leucine-rich repeats and transmembrane domains 2
Gene Aliases
A230084J22, AI841794
Chromosome Location
Chr.6: 119315133 - 119332270 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172492.3 NP_766080.1 1 - 2 274 63
GenBank mRNA AK039006.1 - 1 - 2 108 63
AK045494.1 - 1 - 2 188 63
BC058944.1 - 1 - 2 241 63

View Full Product Details