Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Asprv1
Entrez Gene ID
67855
Gene Name
aspartic peptidase, retroviral-like 1
Gene Aliases
2300003P22Rik, AA986851, SASP, SASPase, Taps
Chromosome Location
Chr.6: 86628174 - 86629710 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026414.2 NP_080690.2 1 - 1 1326 97
GenBank mRNA AK004007.1 - 1 - 1 773 97
AK160494.1 - 1 - 1 1267 97
BC057938.1 - 1 - 1 1339 97
BC108357.1 - 1 - 1 1224 97
BC119099.1 - 1 - 1 842 97
BC119101.1 - 1 - 1 842 97
DQ841260.1 - 1 - 1 1326 97

View Full Product Details