The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199307.2 | NP_955011.1 | 14 - 15 | 1749 | 69 |
GenBank mRNA | AK134088.1 | - | 14 - 15 | 1750 | 69 |
AK163790.1 | - | 11 - 12 | 1662 | 69 | |
AK197559.1 | - | 1 - 2 | 80 | 69 | |
BC038057.1 | - | 5 - 6 | 563 | 69 | |
BC060648.1 | - | 14 - 15 | 1689 | 69 |