Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptch2
Entrez Gene ID
19207
Gene Name
patched homolog 2
Gene Aliases
ptc2
Chromosome Location
Chr.4: 117095785 - 117115383 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008958.2 NP_032984.1 3 - 4 457 73
GenBank mRNA AB010833.1 - 3 - 4 457 73
BC058397.1 - 3 - 4 717 73

View Full Product Details