The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011156.2 | NP_035286.1 | 7 - 8 | 941 | 96 |
GenBank mRNA | AB007631.1 | - | 7 - 8 | 824 | 96 |
AK053816.1 | - | 7 - 8 | 952 | 96 | |
AK076257.1 | - | 7 - 8 | 940 | 96 | |
AK133875.1 | - | 7 - 8 | 940 | 96 | |
AK141851.1 | - | 7 - 8 | 950 | 96 | |
AK161528.1 | - | 7 - 8 | 940 | 96 | |
AK163787.1 | - | 7 - 8 | 953 | 96 | |
AK170570.1 | - | 7 - 8 | 950 | 96 | |
BC012869.1 | - | 7 - 8 | 941 | 96 | |
BC050830.1 | - | 7 - 8 | 934 | 96 |