Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rab42
Entrez Gene ID
242681
Gene Name
RAB42, member RAS oncogene family
Gene Aliases
9530096D07Rik, Rab42-ps
Chromosome Location
Chr.4: 132300438 - 132303403 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081651.1 NP_001075120.1 1 - 2 245 90
GenBank mRNA AB232642.1 - 1 - 2 226 90
BC132060.1 - 1 - 2 274 90
BC132062.1 - 1 - 2 274 90

View Full Product Details