Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Selm
Entrez Gene ID
114679
Gene Name
selenoprotein M
Gene Aliases
1500040L08Rik, A230103K18, Sepm
Chromosome Location
Chr.11: 3514702 - 3517351 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BI731890.1 - 2 - 3 412 85

View Full Product Details