Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Smtn
Entrez Gene ID
29856
Gene Name
smoothelin
Gene Aliases
smsmo
Chromosome Location
Chr.11: 3517522 - 3539654 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159284.1 NP_001152756.1 11 - 12 1747 120
NM_001284427.1 NP_001271356.1 10 - 11 1696 120
NM_001284428.1 NP_001271357.1 11 - 12 1804 120
NM_001284429.1 NP_001271358.1 11 - 12 1749 120
NM_013870.3 NP_038898.2 10 - 11 1696 120
GenBank mRNA AF064236.1 - 10 - 11 1643 120
AJ010305.1 - 10 - 11 1643 120
AK030932.1 - 10 - 11 1697 120
AK085412.1 - 11 - 12 1711 120
AK138898.1 - 11 - 12 1761 120
AK160492.1 - 1 - 2 151 120
AK166385.1 - 11 - 12 1764 120
AK170516.1 - 10 - 11 1656 120
BC010599.1 - 10 - 11 1637 120
BC066192.1 - 11 - 12 1622 120
BC069836.1 - 11 - 12 1622 120
BC069839.1 - 11 - 12 1622 120

View Full Product Details