Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Smtn
Entrez Gene ID
29856
Gene Name
smoothelin
Gene Aliases
smsmo
Chromosome Location
Chr.11: 3517522 - 3539654 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159284.1 NP_001152756.1 16 - 17 2382 107
NM_001284427.1 NP_001271356.1 15 - 16 2331 107
NM_001284428.1 NP_001271357.1 16 - 17 2439 107
NM_001284429.1 NP_001271358.1 16 - 17 2384 107
NM_013870.3 NP_038898.2 15 - 16 2331 107
GenBank mRNA AF064236.1 - 15 - 16 2278 107
AJ010305.1 - 15 - 16 2278 107
AK030932.1 - 15 - 16 2332 107
AK083947.1 - 5 - 6 585 107
AK085412.1 - 16 - 17 2347 107
AK138898.1 - 16 - 17 2397 107
AK160492.1 - 6 - 7 786 107
AK166385.1 - 16 - 17 2400 107
AK170516.1 - 15 - 16 2291 107
BC010599.1 - 15 - 16 2272 107
BC066192.1 - 16 - 17 2257 107
BC069836.1 - 16 - 17 2257 107
BC069839.1 - 16 - 17 2257 107

View Full Product Details