Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nf2
Entrez Gene ID
18016
Gene Name
neurofibromatosis 2
Gene Aliases
merlin
Chromosome Location
Chr.11: 4757650 - 4849544 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252250.1 NP_001239179.1 6 - 7 1158 90
NM_001252251.1 NP_001239180.1 6 - 7 1158 90
NM_001252252.1 NP_001239181.1 6 - 7 1158 90
NM_001252253.1 NP_001239182.1 6 - 7 623 90
NM_010898.4 NP_035028.2 6 - 7 1158 90
GenBank mRNA AK017343.1 - 6 - 7 624 90
AK045998.1 - 6 - 7 980 90
AK167685.1 - 3 - 4 249 90
BC005442.1 - 6 - 7 1024 90
DV654856.1 - 3 - 4 216 90
L22987.1 - 6 - 7 616 90
L22988.1 - 6 - 7 616 90
L22989.1 - 6 - 7 616 90
L27090.1 - 6 - 7 609 90
L27105.1 - 6 - 7 609 90
L28176.1 - 6 - 7 916 90
X74671.1 - 6 - 7 1175 90

View Full Product Details