Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myb
Entrez Gene ID
17863
Gene Name
myeloblastosis oncogene
Gene Aliases
AI550390, M16449, c-myb
Chromosome Location
Chr.10: 21124930 - 21160984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198914.1 NP_001185843.1 2 - 3 406 84
NM_010848.3 NP_034978.3 2 - 3 406 84
GenBank mRNA AK088079.1 - 2 - 3 327 84
AK088598.1 - 2 - 3 328 84
AK132250.1 - 2 - 3 271 84
AK137965.1 - 2 - 3 272 84
BC011513.1 - 2 - 3 314 84
M12848.1 - 2 - 3 404 84
X02774.1 - 2 - 3 176 84

View Full Product Details